1,4-Dichlorobenzene ≥99.0%

Supplier: TCI America
Warning

Synonyms: p-Dichlorobenzene

D0687-500G D0687-25G
TCD0687-500GEA 36.93 USD
TCD0687-500G TCD0687-25G
1,4-Dichlorobenzene ≥99.0%
1,4-Dichlorobenzene

CAS Number: 106-46-7
MDL Number: MFCD00000604
Molecular Formula: C6H4Cl2
Molecular Weight: 147.00
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 173
Melting point (°C): 55
Flash Point (°C): 65

Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 173 °C
Melting Pt: 55 °C
Density: 1.458 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000604
CAS Number: 106-46-7
EINECS: 203-400-5
UN: 3077
ADR: 9,III
Merck Index: 13,03082
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1d32210e3174a3458f1bcba6fa72c808b7aaad1&vid=3e3272ffbad58c328b6a31462ae9562c47a41470&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR