3,5-Xylidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,5-Dimethylaniline, 5-Amino-m-xylene

D0671-500ML D0671-25ML
TCD0671-500MLEA 165 USD
TCD0671-500ML TCD0671-25ML
3,5-Xylidine ≥98.0% (by GC, titration analysis)
3,5-Xylidine

CAS Number: 108-69-0
MDL Number: MFCD00007813
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 221
Flash Point (°C): 97
Freezing point (°C): 6
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 3,5-Xylidine

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 221 °C
Melting Pt: 9.8 °C
Density: 0.97 (20 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00007813
CAS Number: 108-69-0
EINECS: 203-607-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23ae08a33605c4d5149064442955d16a88f7b860&vid=f106ab6d6888c595cc88d3d92b1eff469f688fb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR