2,6-Xylidine ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
2-Amino-m-xylene, 2,6-Dimethylaniline
D0669-25ML
D0669-100ML
TCD0669-25MLEA
34.9
USD
TCD0669-25ML
TCD0669-100ML
2,6-Xylidine ≥99.0%
2,6-Xylidine
CAS Number: 87-62-7
MDL Number: MFCD00007747
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 216
Flash Point (°C): 91
Freezing point (°C): 10
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 2,6-Xylidine
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 216 °C Melting Pt: 10…12 °C Density: 0.98 (20 °C) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007747 CAS Number: 87-62-7 EINECS: 201-758-7 UN: 1711 ADR: 6.1,II Merck Index: 12,10217 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...