Formaldehyde dimethyl acetal ≥98.0%

Supplier: TCI America
Danger

Synonyms: Formal, Methylal, Dimethoxymethane

D0637-500ML D0637-25ML
TCD0637-500MLEA 32.4 USD
TCD0637-500ML TCD0637-25ML
Formaldehyde dimethyl acetal ≥98.0%
Formaldehyde dimethyl acetal

CAS Number: 109-87-5
MDL Number: MFCD00008495
Molecular Formula: C3H8O2
Molecular Weight: 76.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 42
Flash Point (°C): -32
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Dimethylformal, Formal, Formaldehyde Dimethyl Acetal, Methylal, Methylene Glycol Dimethyl Ether

Formula: CH₂(OCH₃)₂
Boiling Pt: 42 °C
Melting Pt: -105 °C
Density: 0.87 (20 °C)
Flash Pt: -32 °C
Storage Temperature: Ambient
MDL Number: MFCD00008495
CAS Number: 109-87-5
EINECS: 203-714-2
UN: 1234
ADR: 3,II
Merck Index: 12,06093
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11b29644311497472fab8d7152520936a239dedc&vid=ad6b8d30fdf42b49b1326bd5f49cfda3fd15d9b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR