2,5-Dimethoxyaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 102-56-7
MDL Number: MFCD00008368
Molecular Formula: C8H11NO2
Molecular Weight: 153.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 159
Melting point (°C): 81
Flash Point (°C): 150
Other Product Names / Synonyms: Aminohydroquinone Dimethyl Ether
Formula:
C₈H₁₁NO₂ Boiling Pt: 159 °C Melting Pt: 81 °C Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008368 CAS Number: 102-56-7 EINECS: 203-040-9 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...