2,5-Dimercapto-1,3,4-thiadiazole ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 1072-71-5
MDL Number: MFCD00003103
Molecular Formula: C2H2N2S3
Molecular Weight: 150.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 156
Lambda max.: 320 nm (CH3CN)
Product Categories: Analytical Science, Bipyridyls, etc. (Chelating Agents)
Other Product Names / Synonyms: 2,5-Dimercapto-1,3,4-thiadiazole
Formula:
C₂H₂N₂S₃ Melting Pt: 156 °C |
MDL Number:
MFCD00003103 CAS Number: 1072-71-5 EINECS: 214-014-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...