4-Ethylresorcinol ≥98.0%

Supplier: TCI America
Warning

Synonyms: 4-Ethylbenzene-1,3-diol

D0579-5G D0579-25G
TCD0579-005GEA 56 USD
TCD0579-005G TCD0579-025G
4-Ethylresorcinol ≥98.0%
4-Ethylresorcinol

CAS Number: 2896-60-8
MDL Number: MFCD00002283
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 155
Melting point (°C): 98


Other Product Names / Synonyms: 2,4-Dihydroxyethylbenzene

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 155 °C
Melting Pt: 98 °C
MDL Number: MFCD00002283
CAS Number: 2896-60-8
EINECS: 220-777-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf1696830d6683df97ae0cf860ca655e7fe0d80d&vid=bf1696830d6683df97ae0cf860ca655e7fe0d80d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR