2,4-Dihydroxybenzophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (2,4-Dihydroxyphenyl)(phenyl)methanone, DHBP

D0573-25G D0573-500G
TCD0573-025GEA 23 USD
TCD0573-025G TCD0573-500G
2,4-Dihydroxybenzophenone ≥98.0% (by GC, titration analysis)
2,4-Dihydroxybenzophenone

CAS Number: 131-56-6
MDL Number: MFCD00002277
Molecular Formula: C13H10O3
Molecular Weight: 214.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 144
Flash Point (°C): 234


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: 4-Benzoylresorcinol

Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 391 °C (1013 hPa)
Melting Pt: 144 °C
Density: 0.696 g/cm³ (20 °C)
Flash Pt: 234 °C
Storage Temperature: Ambient
MDL Number: MFCD00002277
CAS Number: 131-56-6
EINECS: 205-029-4
Merck Index: 13,01107
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=838ba15bc8da36d5393f34d684aa2dd20ed65d6c&vid=838ba15bc8da36d5393f34d684aa2dd20ed65d6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR