3,4-Dihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D0566-25G D0566-250G
TCD0566-025GEA 58 USD
TCD0566-025G TCD0566-250G
3,4-Dihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
3,4-Dihydroxybenzaldehyde

CAS Number: 139-85-5
MDL Number: MFCD00003370
Molecular Formula: C7H6O3
Molecular Weight: 138.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 156


Other Product Names / Synonyms: Protocatechualdehyde

Formula: C₇H₆O₃
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 156 °C
Density: 1.505 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003370
CAS Number: 139-85-5
EINECS: 205-377-7
Merck Index: 12,08079
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8417e759632e6ebf07a73190c07bfb2c5cddae9&vid=f842dc85342ccdaa03115d1ada66369b013adc95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR