3,4-Dihydro-2H-pyran ≥97.0%

Supplier: TCI America

D0555-500ML D0555-25ML
TCD0555-500MLEA 123 USD
TCD0555-500ML TCD0555-25ML
3,4-Dihydro-2H-pyran ≥97.0%
3,4-Dihydro-2H-pyran

CAS Number: 110-87-2
MDL Number: MFCD00006558
Molecular Formula: C5H8O
Molecular Weight: 84.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): -6
Specific Gravity (20/20): 0.93


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: DHP

Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 86 °C
Density: 0.93 (20 °C)
Flash Pt: –6 °C
MDL Number: MFCD00006558
CAS Number: 110-87-2
EINECS: 203-810-4
UN: 2376
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce63b35d82bafde9ec26e24d77119ceafa069ecc&vid=ce63b35d82bafde9ec26e24d77119ceafa069ecc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR