Diethyl sulphate ≥98.0%

Supplier: TCI America
Danger

D0525-25ML D0525-500ML
TCD0525-25MLEA 20 USD
TCD0525-25ML TCD0525-500ML
Diethyl sulphate ≥98.0%
Diethyl sulphate

CAS Number: 64-67-5
MDL Number: MFCD00009099
Molecular Formula: C4H10O4S
Molecular Weight: 154.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Melting point (°C): -25
Flash Point (°C): 110
Specific Gravity (20/20): 1.18

Formula: C₄H₁₀O₄S
Boiling Pt: 208 °C
Melting Pt: –25 °C
Density: 1.18 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009099
CAS Number: 64-67-5
EINECS: 200-589-6
UN: 1594
ADR: 6.1,II
Merck Index: 13,03156
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad2bd429f6edbd5958c5b244be4c8c9a52543149&vid=ad2bd429f6edbd5958c5b244be4c8c9a52543149&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR