Bis(2-ethylhexyl)amine ≥98.0%

Supplier: TCI America

D0510-500ML D0510-25ML
TCD0510-500MLEA 68 USD
TCD0510-500ML TCD0510-25ML
Bis(2-ethylhexyl)amine ≥98.0%
Bis(2-ethylhexyl)amine

CAS Number: 106-20-7
MDL Number: MFCD00009489
Molecular Formula: C16H35N
Molecular Weight: 241.46
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 121
Specific Gravity (20/20): 0.80


Other Product Names / Synonyms: Bis(2-ethylhexyl)amine, Dioctylamine

Formula: C₁₆H₃₅N
Boiling Pt: 123 °C
Melting Pt: ? -60 °C
Density: 0.80 (20 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00009489
CAS Number: 106-20-7
EINECS: 203-372-4
UN: 2922
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24eb1ed4617a26fbb4a59f143c4b8b6df933cd38&vid=24eb1ed4617a26fbb4a59f143c4b8b6df933cd38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR