N,N-Diethylformamide ≥99.0%

Supplier: TCI America
Warning

D0506-25ML D0506-100ML
TCD0506-25MLEA 27 USD
TCD0506-25ML TCD0506-100ML
N,N-Diethylformamide ≥99.0%
N,N-Diethylformamide

CAS Number: 617-84-5
MDL Number: MFCD00003287
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 60
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: Formyldiethylamine

Formula: C₅H₁₁NO
Boiling Pt: 177 °C
Density: 0.91 (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00003287
CAS Number: 617-84-5
EINECS: 210-533-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b585ec202c948282af306bb85baaddf12a0385d&vid=4b585ec202c948282af306bb85baaddf12a0385d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR