N,N-Diethylethylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(Diethylamino)ethylamine, 2-Aminoethyldiethylamine

D0505-500ML D0505-25ML
TCD0505-500MLEA 136 USD
TCD0505-500ML TCD0505-25ML
N,N-Diethylethylenediamine ≥98.0% (by GC, titration analysis)
N,N-Diethylethylenediamine

CAS Number: 100-36-7
MDL Number: MFCD00008176
Molecular Formula: C6H16N2
Molecular Weight: 116.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 40
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: 2-Diethylaminoethylamine

Formula: C₆H₁₆N₂
MW: 116.21 g/mol
Boiling Pt: 146 °C
Melting Pt: –70 °C
Density: 0.82 (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00008176
CAS Number: 100-36-7
EINECS: 202-844-7
UN: 2685
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e428e7cdd3c8d6a47a712233910569b782c259&vid=a0e428e7cdd3c8d6a47a712233910569b782c259&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR