Diethylenetriamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
2,2'-Iminodi(ethylamine), DIEN, Diethylenetriamine
D0493-500ML
D0493-25ML
TCD0493-500MLEA
53.92
USD
TCD0493-500ML
TCD0493-25ML
Diethylenetriamine ≥98.0% (by GC, titration analysis)
Diethylenetriamine
CAS Number: 111-40-0
MDL Number: MFCD00008171
Molecular Formula: C4H13N3
Molecular Weight: 103.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 209
Melting point (°C): -35
Flash Point (°C): 97
Specific Gravity (20/20): 0.95
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Monomers, Bioscience, Polyamines
Other Product Names / Synonyms: 2,2'-Diaminodiethylamine
Formula:
C₄H₁₃N₃ MW: 103.17 g/mol Boiling Pt: 209 °C Melting Pt: –35 °C Density: 0.95 (20 °C) Flash Pt: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008171 CAS Number: 111-40-0 EINECS: 203-865-4 UN: 2079 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...