N,N-Diethylaniline ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

D0475-25ML D0475-500ML
TCD0475-25MLEA 17.27 USD
TCD0475-25ML TCD0475-500ML
N,N-Diethylaniline ≥99.0% (by GC, titration analysis)
N,N-Diethylaniline

CAS Number: 91-66-7
MDL Number: MFCD00009042
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 216
Melting point (°C): -38
Flash Point (°C): 92
Specific Gravity (20/20): 0.93

Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 216 °C
Melting Pt: –38 °C
Density: 0.93 (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00009042
CAS Number: 91-66-7
EINECS: 202-088-8
UN: 2432
ADR: 6.1,III
Merck Index: 13,03140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b93b00d7f5488f6746219789d122490d0d267959&vid=e0d210652a8e75275d5b408285d82096391d9284&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR