Formaldehyde diethyl acetal ≥99.0%
Supplier: TCI America
Synonyms:
Ethylal, Diethoxymethane
D0457-500ML
D0457-25ML
TCD0457-500MLEA
68.16
USD
TCD0457-500ML
TCD0457-25ML
Formaldehyde diethyl acetal ≥99.0%
Formaldehyde diethyl acetal
CAS Number: 462-95-3
MDL Number: MFCD00009246
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 21
Specific Gravity (20/20): 0.83
Other Product Names / Synonyms: Diethoxymethane, Diethylformal, Formaldehyde Diethyl Acetal, Methylene Glycol Diethyl Ether
Formula:
C₅H₁₂O₂ MW: 104.15 g/mol Boiling Pt: 85 °C Melting Pt: -66 °C Density: 0.83 (20 °C) Flash Pt: 21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009246 CAS Number: 462-95-3 EINECS: 207-330-6 UN: 2373 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...