Acetal ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1,1-Diethoxyethane, Acetaldehyde diethyl acetal
D0455-500ML
D0455-25ML
TCD0455-500MLEA
91.88
USD
TCD0455-500ML
TCD0455-25ML
Acetal ≥98.0%
Acetal
CAS Number: 105-57-7
MDL Number: MFCD00009243
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Melting point (°C): -100
Flash Point (°C): -21
Specific Gravity (20/20): 0.83
Other Product Names / Synonyms: Acetaldehyde Diethyl Acetal, 1,1-Diethoxyethane, Diethyl Acetal, Ethylidene Diethyl Ether
Formula:
C₆H₁₄O₂ Boiling Pt: 103 °C Melting Pt: –100 °C Density: 0.83 (20 °C) Flash Pt: –21 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009243 CAS Number: 105-57-7 EINECS: 203-310-6 UN: 1088 ADR: 3,II Merck Index: 13,00040 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...