Diethoxy(dimethyl)silane ≥98.0%

Supplier: TCI America

Synonyms: Dimethyldiethoxysilane

D0454-25ML D0454-100ML
TCD0454-25MLEA 23 USD
TCD0454-25ML TCD0454-100ML
Diethoxy(dimethyl)silane ≥98.0%
Diethoxy(dimethyl)silane

CAS Number: 78-62-6
MDL Number: MFCD00009068
Molecular Formula: C6H16O2Si
Molecular Weight: 148.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Melting point (°C): -70
Flash Point (°C): 15
Specific Gravity (20/20): 0.84

Formula: C₆H₁₆O₂Si
MW: 148.28 g/mol
Boiling Pt: 114
Melting Pt: -87
Density: 0.84 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00009068
CAS Number: 78-62-6
EINECS: 201-127-6
UN: 2380
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=467a41ff9172774a082a1a6cdb84a81f8ac2963b&vid=467a41ff9172774a082a1a6cdb84a81f8ac2963b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR