Didecylamine ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D0446-5G D0446-25G
TCD0446-005GEA 70.61 USD
TCD0446-005G TCD0446-025G
Didecylamine ≥97.0% (by titrimetric analysis)
Didecylamine

CAS Number: 1120-49-6
MDL Number: MFCD00048396
Molecular Formula: C20H43N
Molecular Weight: 297.57
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Freezing point (°C): 32

Formula: C₂₀H₄₃N
MW: 297.57 g/mol
MDL Number: MFCD00048396
CAS Number: 1120-49-6
EINECS: 214-312-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48d4ce5d44fcb60ae0d818d5c2f5b6535d115792&vid=48d4ce5d44fcb60ae0d818d5c2f5b6535d115792&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR