Dicyclohexylamine ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
DCHA
D0435-500ML
D0435-25ML
TCD0435-500MLEA
33.55
USD
TCD0435-500ML
TCD0435-25ML
Dicyclohexylamine ≥99.0%
Dicyclohexylamine
CAS Number: 101-83-7
MDL Number: MFCD00011658
Molecular Formula: C12H23N
Molecular Weight: 181.32
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 256
Melting point (°C): 0
Flash Point (°C): 105
Specific Gravity (20/20): 0.91
Formula:
C₁₂H₂₃N MW: 181.32 g/mol Boiling Pt: 256 °C Melting Pt: –2 °C Density: 0.91 (20 °C) Flash Pt: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011658 CAS Number: 101-83-7 EINECS: 202-980-7 UN: 2565 ADR: 8,III Merck Index: 13,03122 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...