2,6-Dichlorotoluene ≥99.0%
Supplier: TCI America
|
|
Synonyms:
1,3-Dichloro-2-methylbenzene
D0422-25G
D0422-500G
TCD0422-025GEA
33
USD
TCD0422-025G
TCD0422-500G
2,6-Dichlorotoluene ≥99.0%
2,6-Dichlorotoluene
CAS Number: 118-69-4
MDL Number: MFCD00000576
Molecular Formula: C7H6Cl2
Molecular Weight: 161.03
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 92
Specific Gravity (20/20): 1.27
Formula:
C₇H₆Cl₂ MW: 161.03 g/mol Boiling Pt: 198 °C Melting Pt: 2 °C Density: 1.27 (20 °C) Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000576 CAS Number: 118-69-4 EINECS: 204-269-7 UN: 3082 ADR: 9,III |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...