α,4-Dichlorotoluene ≥98.0%

Supplier: TCI America

Synonyms: 1-Chloro-4-(chloromethyl)benzene, 4-Chlorobenzyl chloride

D0421-500G D0421-25G
TCD0421-500GEA 98 USD
TCD0421-500G TCD0421-25G
α,4-Dichlorotoluene ≥98.0%
α,4-Dichlorotoluene

CAS Number: 104-83-6
MDL Number: MFCD00000914
Molecular Formula: C7H6Cl2
Molecular Weight: 161.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 222
Flash Point (°C): 97
Freezing point (°C): 27


Other Product Names / Synonyms: alpha,4-Dichlorotoluene

Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 222 °C
Melting Pt: 28 - 30 °C
Density: 1.259 g/cm³ (25 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00000914
CAS Number: 104-83-6
EINECS: 203-242-7
UN: 3427
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0c9e39d248d340d96f9f6c34e26ba236bb7b3a2&vid=ebe0f78b81169f82b40bd9b3534c54655368c7cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR