2,6-Dichloropyridine ≥97.0%

Supplier: TCI America
Warning

D0410-25G D0410-500G
TCD0410-025GEA 17 USD
TCD0410-025G TCD0410-500G
2,6-Dichloropyridine ≥97.0%
2,6-Dichloropyridine

CAS Number: 2402-78-0
MDL Number: MFCD00006244
Molecular Formula: C5H3Cl2N
Molecular Weight: 147.99
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 212
Melting point (°C): 86
Flash Point (°C): 106

Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 212 °C
Melting Pt: 86 °C
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00006244
CAS Number: 2402-78-0
EINECS: 219-282-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15c754ead1db824f6e22793ed2fe123e5078c3df&vid=15c754ead1db824f6e22793ed2fe123e5078c3df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR