2,4-Dichlorophenol ≥98.0%

Supplier: TCI America
Danger

Synonyms: 1,3-Dichloro-4-hydroxybenzene, 1-Hydroxy-2,4-dichlorobenzene

D0392-500G D0392-25G
TCD0392-500GEA 367 USD
TCD0392-500G TCD0392-25G
2,4-Dichlorophenol ≥98.0%
2,4-Dichlorophenol

CAS Number: 120-83-2
MDL Number: MFCD00002169
Molecular Formula: C6H4Cl2O
Molecular Weight: 163.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 42
Flash Point (°C): 114


Product Categories: Analytical Science, Chlorinated Compounds (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: 2,4-DCP

Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: 210 °C
Melting Pt: 42 °C
Density: 1.4 g/cm³ (20 °C)
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00002169
CAS Number: 120-83-2
EINECS: 204-429-6
UN: 2928
ADR: 6.1,II
Merck Index: 13,03098
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05d97495682714188f9bd3caf4f3c20729373d3c&vid=2925f69f7ac62a956c52e06f4efb1fa79bf8bac3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR