1,2-Dichloro-3-nitrobenzene ≥99.0%

Supplier: TCI America

Synonyms: 2,3-Dichloronitrobenzene

D0386-500G D0386-25G
TCD0386-500GEA 199 USD
TCD0386-500G TCD0386-25G
1,2-Dichloro-3-nitrobenzene ≥99.0%
1,2-Dichloro-3-nitrobenzene

CAS Number: 3209-22-1
MDL Number: MFCD00007062
Molecular Formula: C6H3Cl2NO2
Molecular Weight: 192.00
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 257
Melting point (°C): 62
Flash Point (°C): 123

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Boiling Pt: 257 °C
Melting Pt: 62 °C
Density: 1.721 g/cm³ (14 °C)
Flash Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00007062
CAS Number: 3209-22-1
EINECS: 221-717-7
UN: 1578
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a928dfd4daeb5f456dbb0bbc0064ceb23734a775&vid=b82ba7bd94d2cd42a40261b0ef6042bb272f2d1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR