Dichlone ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2,3-Dichloro-1,4-naphthoquinone

D0384-500G D0384-25G
TCD0384-500GEA 107.83 USD
TCD0384-500G TCD0384-25G
Dichlone ≥95.0% (by GC, titration analysis)
Dichlone

CAS Number: 117-80-6
MDL Number: MFCD00001677
Molecular Formula: C10H4Cl2O2
Molecular Weight: 227.04
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 196
Flash Point (°C): 190

Formula: C₁₀H₄Cl₂O₂
MW: 227.05 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 196 °C
Density: 1.44 g/cm³ (16 °C)
Flash Pt: 190 °C
MDL Number: MFCD00001677
CAS Number: 117-80-6
EINECS: 204-210-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,03071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=851d209e9e80f32d7e39b3ac41ffb354bbc0a2be&vid=851d209e9e80f32d7e39b3ac41ffb354bbc0a2be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR