1,4-Dichlorobutane ≥98.0%

Supplier: TCI America
Danger

D0349-500ML D0349-25ML
TCD0349-500MLEA 50 USD
TCD0349-500ML TCD0349-25ML
1,4-Dichlorobutane ≥98.0%
1,4-Dichlorobutane

CAS Number: 110-56-5
MDL Number: MFCD00001011
Molecular Formula: C4H8Cl2
Molecular Weight: 127.01
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 53
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: Tetramethylene Chloride

Formula: ClH₂C(CH₂)₂CH₂Cl
MW: 127.01 g/mol
Boiling Pt: 153.9
Melting Pt: -38
Density: 1.14 (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00001011
CAS Number: 110-56-5
EINECS: 203-778-1
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c6768cd3765b893bc71bce7ff54119d98be9568&vid=08390a0e2c5858d1826c1c7261448e71530376f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR