1,3-Dichlorobenzene ≥98.0%

Supplier: TCI America
Warning

D0333-500G D0333-25G
TCD0333-500GEA 76.3 USD
TCD0333-500G TCD0333-25G
1,3-Dichlorobenzene ≥98.0%
1,3-Dichlorobenzene

CAS Number: 541-73-1
MDL Number: MFCD00000573
Molecular Formula: C6H4Cl2
Molecular Weight: 147.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Melting point (°C): -25
Flash Point (°C): 62
Specific Gravity (20/20): 1.29

Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 173
Melting Pt: -25
Density: 1.29 (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000573
CAS Number: 541-73-1
EINECS: 208-792-1
UN: 3082
ADR: 9 III
Merck Index: 13,03080
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=020f37232391574bedb70d4e338868ec0057ed73&vid=e13491800b3afd2a8860e41f5653539367f9339f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR