2,6-Dichlorobenzaldehyde ≥97.0%

Supplier: TCI America
Warning

D0331-25G D0331-100G
TCD0331-25GEA 57 USD
TCD0331-25G TCD0331-100G
2,6-Dichlorobenzaldehyde ≥97.0%
2,6-Dichlorobenzaldehyde

CAS Number: 83-38-5
MDL Number: MFCD00003307
Molecular Formula: C7H4Cl2O
Molecular Weight: 175.01
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 231
Melting point (°C): 71
Flash Point (°C): 100

Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 231 °C
Melting Pt: 71 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003307
CAS Number: 83-38-5
EINECS: 201-472-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04e674fbb986bf9f58af223efcf2ad3ade50c915&vid=543c5c49b10ca87ffb2963a2a46983d9549088e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR