2,4-Dichloroaniline ≥99.0%

Supplier: TCI America
Danger

D0321-500G D0321-25G D0321-100G
TCD0321-500GEA 328 USD
TCD0321-500G TCD0321-25G TCD0321-100G
2,4-Dichloroaniline ≥99.0%
2,4-Dichloroaniline

CAS Number: 554-00-7
MDL Number: MFCD00007661
Molecular Formula: C6H5Cl2N
Molecular Weight: 162.01
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 242
Melting point (°C): 62
Flash Point (°C): 110

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 242 °C
Melting Pt: 62 °C
Density: 1.567 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007661
CAS Number: 554-00-7
EINECS: 209-057-8
UN: 3442
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9173028e44f310b9d3e41bec06641044e39bd89&vid=c07bb21df8ee064de8c87f24c314c6bd0134d874&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR