Methyl dichloroacetate ≥99.0%

Supplier: TCI America
Warning

D0309-500G D0309-25G
TCD0309-500GEA 58 USD
TCD0309-500G TCD0309-25G
Methyl dichloroacetate ≥99.0%
Methyl dichloroacetate

CAS Number: 116-54-1
MDL Number: MFCD00000843
Molecular Formula: C3H4Cl2O2
Molecular Weight: 142.96
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 143
Melting point (°C): -52
Flash Point (°C): 72
Specific Gravity (20/20): 1.38


Other Product Names / Synonyms: Dichloroacetic Acid Methyl Ester

Formula: C₃H₄Cl₂O₂
MW: 142.97 g/mol
Boiling Pt: 143 °C
Melting Pt: -52 °C
Density: 1.38 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00000843
CAS Number: 116-54-1
EINECS: 204-146-8
UN: 2299
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=047457e9ebee77c7595a73cfecb18a77bd2ebdcb&vid=047457e9ebee77c7595a73cfecb18a77bd2ebdcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR