3,5-Diaminobenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D0294-500G D0294-25G D0294-100G
TCD0294-500GEA 556 USD
TCD0294-500G TCD0294-025G TCD0294-100G
3,5-Diaminobenzoic acid ≥98.0% (by HPLC, titration analysis)
3,5-Diaminobenzoic acid

CAS Number: 535-87-5
MDL Number: MFCD00007807
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 228

Formula: C₇H₈N₂O₂
Boiling Pt: 546 °C (1013 hPa)
Melting Pt: 228 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007807
CAS Number: 535-87-5
EINECS: 208-621-0
Merck Index: 13,02997
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=105764260764e4ba79e21262635ac8ab1a186796&vid=5769ee9cfa4c193abe39c48dbddeb5c7fa0ec11c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR