Dibutyltin dichloride ≥97.0%

Supplier: TCI America

Synonyms: Dichlorodibutylstannane, Di-n-butyltin dichloride, DBTC

D0223-25G D0223-100G
TCD0223-25GEA 23.38 USD
TCD0223-25G TCD0223-100G
Dibutyltin dichloride ≥97.0%
Dibutyltin dichloride

CAS Number: 683-18-1
MDL Number: MFCD00000518
Molecular Formula: C8H18Cl2Sn
Molecular Weight: 303.84
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 42
Flash Point (°C): 172


Product Categories: Synthetic Organic Chemistry, Grignard Reagents & Alkyl Metals, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Dibutyldichlorotin

Formula: C₈H₁₈Cl₂Sn
Boiling Pt: 135 °C
Melting Pt: 42 °C
Density: 1.37 to 1.4 g/cm³ (20 °C)
Flash Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00000518
CAS Number: 683-18-1
EINECS: 211-670-0
UN: 2928
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f6ae92c377156b5edb974c6080bd5852786d20a&vid=2f6ae92c377156b5edb974c6080bd5852786d20a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR