Dibutylamine ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: Di-n-butylamine, N-butyl-1-propanamine

D0218-500ML D0218-25ML
TCD0218-500MLEA 50.58 USD
TCD0218-500ML TCD0218-25ML
Dibutylamine ≥99.0% (by GC, titration analysis)
Dibutylamine

CAS Number: 111-92-2
MDL Number: MFCD00009429
Molecular Formula: C8H19N
Molecular Weight: 129.25
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 159
Melting point (°C): -59
Flash Point (°C): 43
Specific Gravity (20/20): 0.76

Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 159 °C
Melting Pt: -59 °C
Density: 0.76 (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00009429
CAS Number: 111-92-2
EINECS: 203-921-8
UN: 2248
ADR: 8,II
Merck Index: 13,03058
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ab1184f23fb3f770d58f45690f3311c0bcfebab&vid=4ab1184f23fb3f770d58f45690f3311c0bcfebab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR