1,5-Dibromopentane ≥98.0%

Supplier: TCI America
Warning

D0198-25G D0198-500G
TCD0198-025GEA 19 USD
TCD0198-025G TCD0198-500G
1,5-Dibromopentane ≥98.0%
1,5-Dibromopentane

CAS Number: 111-24-0
MDL Number: MFCD00000268
Molecular Formula: C5H10Br2
Molecular Weight: 229.94
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 221
Flash Point (°C): 79
Specific Gravity (20/20): 1.70


Other Product Names / Synonyms: Pentamethylene Bromide

Formula: BrH₂C(CH₂)₃CH₂Br
MW: 229.94 g/mol
Boiling Pt: 221 °C
Melting Pt: –39 °C
Density: 1.70 (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000268
CAS Number: 111-24-0
EINECS: 203-849-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c324c2f9ac10f7f475cd0491f91ee397f261803&vid=cb43642e15dc50aeaf766396e994cab610caf524&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR