1,3-Dibromobenzene ≥97.0%

Supplier: TCI America
Warning

D0169-25G D0169-500G D0169-100G
TCD0169-025GEA 25 USD
TCD0169-025G TCD0169-500G 76218-950
1,3-Dibromobenzene ≥97.0%
1,3-Dibromobenzene

CAS Number: 108-36-1
MDL Number: MFCD00000078
Molecular Formula: C6H4Br2
Molecular Weight: 235.91
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 219
Melting point (°C): -7
Flash Point (°C): 94
Specific Gravity (20/20): 1.96

Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 218
Melting Pt: -7
Density: 1.96 (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00000078
CAS Number: 108-36-1
EINECS: 203-574-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cedf774ec1fa5c877b6939b2ffe0447b0a9f2f74&vid=3946573d879897b33d8463c885bcdbf07b773358&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR