Dibenzothiophene ≥98.0%
Supplier: TCI America
Synonyms:
Dibenzo[b,d]thiophene
D0148-5G
D0148-25G
TCD0148-005GEA
28
USD
TCD0148-005G
TCD0148-025G
Dibenzothiophene ≥98.0%
Dibenzothiophene
CAS Number: 132-65-0
MDL Number: MFCD00004969
Molecular Formula: C12H8S
Molecular Weight: 184.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 333
Melting point (°C): 99
Flash Point (°C): 170
Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: Dibenzthiophene, Diphenylene Sulfide
Formula:
C₁₂H₈S MW: 184.26 g/mol Boiling Pt: 333 °C Melting Pt: 99 °C Density: 1.1295 g/cm³ (125 °C) Flash Pt: 170 °C |
MDL Number:
MFCD00004969 CAS Number: 132-65-0 EINECS: 205-072-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...