Dibenzofuran ≥98.0%

Supplier: TCI America

D0147-25G D0147-500G
TCD0147-025GEA 26.46 USD
TCD0147-025G TCD0147-500G
Dibenzofuran ≥98.0%
Dibenzofuran

CAS Number: 132-64-9
MDL Number: MFCD00004968
Molecular Formula: C12H8O
Molecular Weight: 168.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 288
Melting point (°C): 83
Flash Point (°C): 130


Other Product Names / Synonyms: Diphenylene Oxide

Formula: C₁₂H₈O
Boiling Pt: 288 °C
Melting Pt: 83 °C
Density: 0.966 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004968
CAS Number: 132-64-9
EINECS: 205-071-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1227945463ecd35852e58ae10ef8786b0f71128&vid=d1227945463ecd35852e58ae10ef8786b0f71128&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR