1,3-Diaminopropane ≥98.0%
Supplier: TCI America
Synonyms:
1,3-Propanediamine, Trimethylenediamine
D0114-25ML
D0114-500ML
TCD0114-025MLEA
24.42
USD
TCD0114-025ML
TCD0114-500ML
1,3-Diaminopropane ≥98.0%
1,3-Diaminopropane
CAS Number: 109-76-2
MDL Number: MFCD00008228
Molecular Formula: C3H10N2
Molecular Weight: 74.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 133
Flash Point (°C): 49
Freezing point (°C): -12
Specific Gravity (20/20): 0.89
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Propanediamine, Trimethylenediamine
Formula:
C₃H₁₀N₂ MW: 74.13 g/mol Boiling Pt: 133 °C Melting Pt: –12 °C Density: 0.89 (20 °C) Flash Pt: 49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008228 CAS Number: 109-76-2 EINECS: 203-702-7 UN: 2734 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...