1,5-Diaminonaphthalene ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1,5-naphthylenediamine, 1,5-Naphthalenediamine
D0101-25G
D0101-500G
D0101-100G
TCD0101-025GEA
77.01
USD
TCD0101-025G
TCD0101-500G
TCD0101-100G
1,5-Diaminonaphthalene ≥98.0% (by GC, titration analysis)
1,5-Diaminonaphthalene
CAS Number: 2243-62-1
MDL Number: MFCD00004029
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 187
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 1,5-Naphthalenediamine
Formula:
C₁₀H₁₀N₂ MW: 158.2 g/mol Boiling Pt: 210 °C Melting Pt: 187 °C Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004029 CAS Number: 2243-62-1 EINECS: 218-817-8 UN: 3077 ADR: 9,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...