1,6-Diaminohexane ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 1,6-Hexanediamine, Hexamethylenediamine

D0095-500G D0095-25G
TCD0095-500GEA 97.94 USD
TCD0095-500G TCD0095-25G
1,6-Diaminohexane ≥99.0% (by GC, titration analysis)
1,6-Diaminohexane

CAS Number: 124-09-4
MDL Number: MFCD00008243
Molecular Formula: C6H16N2
Molecular Weight: 116.21
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 205
Melting point (°C): 42
Flash Point (°C): 85


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Hexamethylenediamine, 1,6-Hexanediamine

Formula: C₆H₁₆N₂
MW: 116.21 g/mol
Boiling Pt: 205 °C
Melting Pt: 42 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00008243
CAS Number: 124-09-4
EINECS: 204-679-6
UN: 2280
ADR: 8,III
Merck Index: 13,04713
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25fe2af8c81dd0a9715db7a8b2c4a7c393240c5c&vid=25fe2af8c81dd0a9715db7a8b2c4a7c393240c5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR