3,3'-Iminobis(propylamine) ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0090-500ML D0090-25ML
TCD0090-500MLEA 127.92 USD
TCD0090-500ML TCD0090-25ML
3,3'-Iminobis(propylamine) ≥98.0% (by GC, titration analysis)
3,3'-Iminobis(propylamine)

CAS Number: 56-18-8
MDL Number: MFCD00008214
Molecular Formula: C6H17N3
Molecular Weight: 131.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 115
Melting point (°C): -14
Flash Point (°C): 117
Specific Gravity (20/20): 0.93


Product Categories: Materials Chemistry, Monomers, Bioscience, Polyamines
Other Product Names / Synonyms: Dipropylenetriamine, 3,3'-Iminobis(propylamine), 3,3'-Iminodipropylamine

Formula: C₆H₁₇N₃
Boiling Pt: 115 °C
Melting Pt: –14 °C
Density: 0.93 (20 °C)
Flash Pt: 117 °C
MDL Number: MFCD00008214
CAS Number: 56-18-8
EINECS: 200-261-2
UN: 2269
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f9ee63eb0a610c9b554aa9a85a77463448a0480&vid=1f9ee63eb0a610c9b554aa9a85a77463448a0480&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR