Diallylamine ≥98.0%

Supplier: TCI America

D0069-500ML D0069-25ML
TCD0069-500MLEA 108 USD
TCD0069-500ML TCD0069-25ML
Diallylamine ≥98.0%
Diallylamine

CAS Number: 124-02-7
MDL Number: MFCD00008642
Molecular Formula: C6H11N
Molecular Weight: 97.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Melting point (°C): -88
Flash Point (°C): 14
Specific Gravity (20/20): 0.79

Formula: C₆H₁₁N
MW: 97.16 g/mol
Boiling Pt: 112 °C
Melting Pt: –88 °C
Density: 0.79 (20 °C)
Flash Pt: 14 °C
MDL Number: MFCD00008642
CAS Number: 124-02-7
EINECS: 204-671-2
UN: 2359
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79b907d0d508a96759d25cabc4381bffb1846cbf&vid=79b907d0d508a96759d25cabc4381bffb1846cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR