Caprinaldehyde ≥97.0%

Supplier: TCI America

D0032-25ML D0032-500ML
TCD0032-025MLEA 27.93 USD
TCD0032-025ML TCD0032-500ML
Caprinaldehyde ≥97.0%
Caprinaldehyde

CAS Number: 112-31-2
MDL Number: MFCD00007031
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Melting point (°C): -5
Flash Point (°C): 94
Specific Gravity (20/20): 0.83
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Decanaldehyde, Decyl Aldehyde

Formula: C₁₀H₂₀O
Boiling Pt: 208 °C
Melting Pt: -5 °C
Density: 0.83 (20 °C)
Flash Pt: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007031
CAS Number: 112-31-2
EINECS: 203-957-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c72fe8d4c64164da5416bf899deccdc7c40f7c2e&vid=5091177eb02c783811e2a394a7a2e8632728124c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR