1-Decanol ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1-Hydroxy decane, 1-Decyl alcohol, Capric alcohol, Decan-1-ol, Nonyl carbinol, Decyl alcohol
D0031-25ML
D0031-500ML
TCD0031-025MLEA
25.09
USD
TCD0031-025ML
TCD0031-500ML
1-Decanol ≥98.0%
1-Decanol
CAS Number: 112-30-1
MDL Number: MFCD00004747
Molecular Formula: C10H22O
Molecular Weight: 158.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 233
Flash Point (°C): 117
Freezing point (°C): 7
Specific Gravity (20/20): 0.83
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Other Plant Hormones
Other Product Names / Synonyms: Decyl Alcohol
Formula:
H₃C(CH₂)₈CH₂OH MW: 158.28 g/mol Boiling Pt: 233 °C Melting Pt: 7 °C Density: 0.83 (20 °C) Flash Pt: 117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004747 CAS Number: 112-30-1 EINECS: 203-956-9 UN: 3082 ADR: 9 III Merck Index: 13,02875 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...