1-Decanethiol ≥95.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
n-Decanethiol, 1-Mercaptodecane, n-Decyl mercaptan
D0016-500ML
D0016-25ML
D0016-100ML
TCD0016-500MLEA
201
USD
TCD0016-500ML
TCD0016-025ML
TCD0016-100ML
1-Decanethiol ≥95.0% (by GC, titration analysis)
1-Decanethiol
CAS Number: 143-10-2
MDL Number: MFCD00004884
Molecular Formula: C10H22S
Molecular Weight: 174.35
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 241
Melting point (°C): -26
Flash Point (°C): 98
Specific Gravity (20/20): 0.84
Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Decyl Mercaptan
Formula:
C₁₀H₂₂S MW: 174.35 g/mol Boiling Pt: 241 °C Melting Pt: –26 °C Density: 0.84 (20 °C) Flash Pt: 98 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004884 CAS Number: 143-10-2 EINECS: 205-584-2 UN: 3334 ADR: 9, |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...