N,N-Dimethyldodecylamine ≥96.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
CAS Number: 112-18-5
MDL Number: MFCD00008970
Molecular Formula: C14H31N
Molecular Weight: 213.41
Purity/Analysis Method: >96.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 149
Melting point (°C): -20
Flash Point (°C): 130
Specific Gravity (20/20): 0.79
Other Product Names / Synonyms: N,N-Dimethyllaurylamine, 1-(Dimethylamino)dodecane
Formula:
C₁₄H₃₁N Boiling Pt: 149 °C Melting Pt: –20 °C Density: 0.79 (20 °C) Flash Pt: 130 °C |
MDL Number:
MFCD00008970 CAS Number: 112-18-5 EINECS: 203-943-8 UN: 2735 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...