Bis(4-nitrophenyl)carbonate ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: Bis(4-nitrophenyl) carbonate

C1481-5G C1481-25G
TCC1481-005GEA 63.8 USD
TCC1481-005G TCC1481-025G
Bis(4-nitrophenyl)carbonate ≥97.0% (by HPLC)
Bis(4-nitrophenyl)carbonate

CAS Number: 5070-13-3
MDL Number: MFCD00007322
Molecular Formula: C13H8N2O7
Molecular Weight: 304.21
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 142


Product Categories: Synthetic Organic Chemistry, Phosgene & Thiophosgene Alternatives
Other Product Names / Synonyms: Carbonic Acid Bis(4-nitrophenyl) Ester

Formula: C₁₃H₈N₂O₇
MW: 304.22 g/mol
Melting Pt: 142 °C
MDL Number: MFCD00007322
CAS Number: 5070-13-3
EINECS: 225-775-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77a7ae9563a0de0dd6b3d11f3977c8b46745e4e3&vid=77a7ae9563a0de0dd6b3d11f3977c8b46745e4e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR