3-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-(3-Chlorophenyl)hydrazine hydrochloride, 3-Chlorophenylhydrazine hydrochloride

C1477-25G C1477-5G
TCC1477-025GEA 79 USD
TCC1477-025G TCC1477-5G
3-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)
3-Chlorophenylhydrazinium chloride

CAS Number: 2312-23-4
MDL Number: MFCD00012935
Molecular Formula: C6H7ClN2
Molecular Weight: 179.04
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₆H₈Cl₂N₂
MW: 179.05 g/mol
MDL Number: MFCD02656655
CAS Number: 2312-23-4
EINECS: 219-005-6

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR